Invitrogen
Manufacturer Code:MA537759
Catalog # PIMA537759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1833CT856.7.20 |
Immunogen | recombinant protein of human DKK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dickkopf 2; dickkopf 2 dickkopf 2 homolog Dickkopf gene 2 dickkopf homolog 1 dickkopf homolog 2 dickkopf homolog 2 (Xenopus laevis) dickkopf homolog 4 dickkopf related protein-2 dickkopf WNT signaling pathway inhibitor 2 dickkopf-2 Dickkopf-related protein 2 Dkk2 dkk-2 Dkk4 hDkk-2 mDkk-2 UNQ682/PRO1316; dickkopf 2 homolog; dickkopf homolog 1; dickkopf homolog 2; dickkopf related protein-2; Dickkopf-2; Dickkopf-related protein 2; dkk-2; hDkk-2; mDkk-2
Gene Aliases: DKK-2; DKK2; UNQ682/PRO1316
UniProt ID: (Human) Q9UBU2, (Mouse) Q9QYZ8
Entrez Gene ID: (Human) 27123, (Mouse) 56811
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.