Invitrogen
Manufacturer Code:PA547318
Catalog # PIPA547318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Dkk-2 Ser26-Ile259 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: dickkopf 2; dickkopf homolog 1; Dickkopf-2; Dickkopf-related protein 2; dkk-2; DKK2 dickkopf homolog 2 (Xenopus laevis) DKK-2 Dickkopf gene 2 dickkopf (Xenopus laevis) homolog 2 dickkopf 2 dickkopf homolog 2 dickkopf related protein-2; mDkk-2
Gene Aliases: Dkk2
UniProt ID: (Mouse) Q9QYZ8
Entrez Gene ID: (Mouse) 56811
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.