Invitrogen
Manufacturer Code:PA511617
Catalog # PIPA511617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 225-251 amino acids from the C-terminal region of human DKK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dickkopf 2 homolog; dickkopf homolog 2; Dickkopf related protein-2; Dickkopf-2; Dickkopf-related protein 2; hDkk-2
Gene Aliases: DKK-2; DKK2; UNQ682/PRO1316
UniProt ID: (Human) Q9UBU2
Entrez Gene ID: (Human) 27123
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.