Proteintech
Manufacturer Code:210511AP150UL
Catalog # 50-173-3230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DKK2 Fusion Protein Ag12405 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dickkopf 2; Dickkopf 2 Dickkopf related protein 2 DKK 2 DKK2 hDkk 2; dickkopf 2 homolog; dickkopf homolog 1; dickkopf homolog 2; dickkopf homolog 4; dickkopf related protein-2; Dickkopf-2; Dickkopf-related protein 2; dkk-2; hDkk-2; mDkk-2
Gene Aliases: DKK-2; DKK2; Dkk4; UNQ682/PRO1316
UniProt ID: (Human) Q9UBU2, (Mouse) Q9QYZ8
Entrez Gene ID: (Human) 27123, (Mouse) 56811, (Rat) 295445
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.