Novus Biologicals
Manufacturer Code:NBP238312
Catalog # NBP238312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: YYKKFSIPDLDRHQLPLDDALLSFAHANCTLIISYQKPKEVVVAESELQKELKKVKTAHSNDGDCK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deleted in a mouse model of primary ciliary dyskinesia; deleted in a mouse model of primary ciliary dyskinesia deleted in primary ciliary dyskinesia homolog (mouse) DKFZP566F084 protein DPCD RP11-529I10.4; Protein DPCD; RP11-529I10.4
Gene Aliases: DPCD
UniProt ID: (Human) Q9BVM2
Entrez Gene ID: (Human) 25911
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.