Invitrogen
Manufacturer Code:PA5101068
Catalog # PIPA5101068
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Dyskerin corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC068171 CBF5 CBF5 homolog DKC Dkc1 DKCX dyskeratosis congenita 1 dyskerin dyskeratosis congenita 1 dyskerin homolog Dyskerin dyskerin pseudouridine synthase 1 H/ACA ribonucleoprotein complex subunit 4 H/ACA ribonucleoprotein complex subunit DKC1 Nap57 NOLA4 Nopp140-associated protein of 57 kDa nucleolar protein family A member 4 Nucleolar protein NAP57 snoRNP protein DKC1 XAP101; CBF5 homolog; dyskeratosis congenita 1, dyskerin; dyskeratosis congenita 1, dyskerin homolog; Dyskerin; H/ACA ribonucleoprotein complex subunit 4; H/ACA ribonucleoprotein complex subunit DKC1; Nopp140-associated protein of 57 kDa; Nucleolar protein family A member 4; Nucleolar protein NAP57; snoRNP protein DKC1
Gene Aliases: BC068171; CBF5; DKC; DKC1; DKCX; NAP57; NOLA4; XAP101
UniProt ID: (Human) O60832, (Rat) P40615, (Mouse) Q9ESX5
Entrez Gene ID: (Human) 1736, (Rat) 170944, (Mouse) 245474
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.