Invitrogen
Manufacturer Code:PA586492
Catalog # PIPA586492
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 50-97 of Human PARK7 / DJ-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Contraception-associated protein 1; DJ-1; epididymis secretory sperm binding protein Li 67p; Fertility protein SP22; Maillard deglycase; Oncogene DJ1; Parkinson disease (autosomal recessive, early onset) 7; Parkinson disease protein 7; Parkinson disease protein 7 homolog; parkinson protein 7; Parkinsonism-associated deglycase; Protein CAP1; Protein DJ-1; Protein DJ-1 Oncogene DJ1 Parkinson disease protein 7 PARK7; Protein/nucleic acid deglycase DJ-1
Gene Aliases: CAP1; DJ-1; DJ1; HEL-S-67p; PARK7; SP22
UniProt ID: (Human) Q99497, (Rat) O88767
Entrez Gene ID: (Human) 11315, (Rat) 117287
Molecular Function: RNA binding protein cysteine protease hydrolase nucleic acid binding protease transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.