Invitrogen
Manufacturer Code:PA572638
Catalog # PIPA572638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 120-149 amino acids from the C-terminal region of DANRE park7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DJ-1 protein; EMBL:AAH834751}; Maillard deglycase; parkinson disease (autosomal recessive, early onset) 7; Parkinson disease protein 7 homolog; Parkinsonism-associated deglycase; protein deglycase DJ-1zDJ-1; protein DJ-1; Protein DJ-1 zDJ-1 34-- Parkinson disease protein 7 homolog park7 {ECO:0000312; Protein DJ-1zDJ-1; Protein/nucleic acid deglycase DJ-1; zDJ-1
Gene Aliases: dj1; park7; zgc:103725
UniProt ID: (Zebrafish) Q5XJ36
Entrez Gene ID: (Zebrafish) 449674
Molecular Function:
RNA binding protein
cysteine protease
hydrolase
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.