Invitrogen
Manufacturer Code:MA526153
Catalog # PIMA526153
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6B6 |
Immunogen | Full length human recombit protein of DIXDC1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Coiled-coil protein DIX1; Coiled-coil-DIX1; DIX domain-containing protein 1; DIXDC1 DIX domain containing 1 CCD1 KIAA1735; Dixin
Gene Aliases: CCD1; DIXDC1; EGK_06890; KIAA1735
UniProt ID: (Human) Q8IVY4
Entrez Gene ID: (Human) 85458, (Rhesus monkey) 710847
Molecular Function:
enzyme modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.