Proteintech
Manufacturer Code:138161AP150UL
Catalog # 50-172-9023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DIXDC1 Fusion Protein Ag4904 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCD1 Coiled coil DIX1 Coiled coil protein DIX1 DIX domain containing 1 DIXDC1 Dixin KIAA1735; Coiled-coil protein DIX1; Coiled-coil-DIX1; DIX domain-containing protein 1; Dixin
Gene Aliases: 4930563F16Rik; BC048182; CCD1; Ccd1Aalpha1L; Ccd1Abeta1L; Ccd1BalphaL; Ccd1BbetaL; Ccd1CL; DIXDC1; KIAA1735; mKIAA1735
UniProt ID: (Human) Q8IVY4, (Mouse) Q80Y83, (Rat) Q2VUH7
Entrez Gene ID: (Human) 85458, (Mouse) 330938, (Rat) 363062
Molecular Function: enzyme modulator signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.