Invitrogen
Manufacturer Code:PA572744
Catalog # PIPA572744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DISP1 antibody for a 16 amino acid synthetic peptide near the amino terminus of human DISP1. The immunogen is within amino acids 20 - 70 of DISP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DISP1 DISPA MGC13130 MGC16796 DKFZP434I0428 dispatched A DISP1 dispatched homolog 1 dispatched homolog 1 (Drosophila) FLJ43740 MGC104180; dispatched A; dispatched homolog 1; Mdispa; Protein dispatched homolog 1; twelve-pass transmembrane protein
Gene Aliases: 1190008H24Rik; Disp; DISP1; DISPA; Icb; Icbins
UniProt ID: (Human) Q96F81, (Mouse) Q3TDN0
Entrez Gene ID: (Human) 84976, (Mouse) 68897
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.