Invitrogen
Manufacturer Code:PA543505
Catalog # PIPA543505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human DISP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chameleon; dispatched 1; dispatched A; dispatched homolog 1; Dispatched homolog 1 (Drosophila) DISPA DKFZP434I0428 FLJ43740 MGC104180 MGC13130 MGC16796; helix; hlx; Mdispa; multipass transmembrane protein; Disp1; Protein chameleon; Protein dispatched homolog 1; protein dispatched homolog 1 pseudogene; twelve-pass transmembrane protein
Gene Aliases: 1190008H24Rik; con; Disp; DISP1; DISPA; Icb; Icbins; zgc:111866
UniProt ID: (Human) Q96F81, (Mouse) Q3TDN0, (Zebrafish) Q6R5J2
Entrez Gene ID: (Human) 84976, (Rabbit) 100350276, (Horse) 100049844, (Pig) 106505093, (Rat) 289338, (Mouse) 68897, (Zebrafish) 619201
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.