Invitrogen
Manufacturer Code:PA518257
Catalog # PIPA518257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SELSGESLLIKTL) corresponding to the C-terminus amino acids of DISP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DISP1 DISPA MGC13130 MGC16796 DKFZP434I0428 dispatched A DISP1 dispatched homolog 1 dispatched homolog 1 (Drosophila) FLJ43740 MGC104180; dispatched A; dispatched homolog 1; Protein dispatched homolog 1
Gene Aliases: DISP1; DISPA
UniProt ID: (Human) Q96F81
Entrez Gene ID: (Human) 84976
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.