Novus Biologicals
Manufacturer Code:NBP281818
Catalog # NB017129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DISP1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human DISP1. The immunogen is located within amino acids 20 - 70 of DISP1. Amino Acid Squence: CQLTPKEATRTKVSPNG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DISPAFLJ43740 dispatched A dispatched homolog 1 (Drosophila) DKFZp434I0428 MGC104180 MGC13130 MGC16796 protein dispatched homolog 1; dispatched A; dispatched homolog 1; Protein dispatched homolog 1
Gene Aliases: DISP1; DISPA
UniProt ID: (Human) Q96F81
Entrez Gene ID: (Human) 84976
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.