R&D Systems
Manufacturer Code:AF3549
Catalog # AF3549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human DISP1 Glu212-Lys492 Ser746-Thr984 Accession # NP_116279 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: DISPA DISPAFLJ43740 Dispatched dispatched A dispatched homolog 1 (Drosophila) DKFZp434I0428 MGC104180 MGC13130 MGC16796 protein dispatched homolog 1; dispatched A; dispatched homolog 1; Protein dispatched homolog 1
Gene Aliases: DISP1; DISPA
UniProt ID: (Human) Q96F81
Entrez Gene ID: (Human) 84976
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.