Proteintech
Manufacturer Code:155001AP150UL
Catalog # 50-173-0272
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf136 DISC1 DISC1 long disrupted in schizophrenia 1 KIAA0457 SCZD9; Disrupted in schizophrenia 1 homolog; Disrupted in schizophrenia 1 protein
Gene Aliases: C1orf136; DISC1; KIAA0457; SCZD9
UniProt ID: (Human) Q9NRI5, (Mouse) Q811T9
Entrez Gene ID: (Human) 27185, (Mouse) 244667
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.