Novus Biologicals
Manufacturer Code:NBP237316
Catalog # NBP237316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6C7B2 |
Immunogen | Purified recombinant fragment of human DIS3L2 (AA: 27-250) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DIS3 mitotic control homolog (S. cerevisiae)-like 2 DIS3-like exonuclease 2 FAM6A member A; DIS3 mitotic control homolog-like 2; DIS3-like exonuclease 2; DIS3L2; family with sequence similarity 6, member A; hDIS3L2
Gene Aliases: DIS3L2; FAM6A; hDIS3L2; PRLMNS
UniProt ID: (Human) Q8IYB7
Entrez Gene ID: (Human) 129563
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.