Proteintech
Manufacturer Code:257921AP150UL
Catalog # 50-173-5072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DIS3L2 Fusion Protein Ag22963 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DIS3 like exonuclease 2 DIS3L2 FAM6A; DIS3 mitotic control homolog-like 2; DIS3-like exonuclease 2; DIS3L2; family with sequence similarity 6, member A; hDIS3L2
Gene Aliases: 4930429A22Rik; 8030493P09Rik; DIS3L2; FAM6A; hDIS3L2; PRLMNS
UniProt ID: (Human) Q8IYB7, (Mouse) Q8CI75
Entrez Gene ID: (Human) 129563, (Mouse) 208718
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.