Novus Biologicals
Manufacturer Code:NBP185209
Catalog # NBP185209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:EAYILFVRKNAIVVLIPKYGLEGTVFFEEKDKPNPQLIYDDEIPSLKIEDTVFHVFDKVKVKIMLDSSNLQHQKIRMSLVEPQIPGISIPTDTSN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810028N01Rik DIS3 mitotic control homolog (S. cerevisiae) dis3p DKFZp667L1817 EC 3.1.13 EC 3.1.13.- EC 3.1.26.- EXOSC11 exosome complex exonuclease RRP44 exosome component 11 FLJ10484 KIAA1008RP11-342J4.3 MGC33035 mitotic control protein dis3 homolog Protein DIS3 homolog Ribosomal RNA-processing protein 44 RRP44bA555G22.1; DIS3 exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 mitotic control homolog; Exosome complex exonuclease RRP44; exosome component 11; mitotic control protein dis3 homolog; Protein DIS3 homolog; Ribosomal RNA-processing protein 44
Gene Aliases: 2810028N01Rik; DIS3; dis3p; EXOSC11; KIAA1008; RRP44
UniProt ID: (Human) Q9Y2L1
Entrez Gene ID: (Human) 22894
Molecular Function: RNA binding protein endoribonuclease exoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.