Proteintech
Manufacturer Code:146891AP150UL
Catalog # 50-172-9643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DIS3 Fusion Protein Ag6117 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DIS3 dis3p EXOSC11 KIAA1008 Protein DIS3 homolog RP11 342J4.3 RRP44; DIS3 exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 mitotic control homolog; Exosome complex exonuclease RRP44; exosome component 11; mitotic control protein dis3 homolog; Protein DIS3 homolog; Ribosomal RNA-processing protein 44
Gene Aliases: 2810028N01Rik; DIS3; dis3p; EXOSC11; KIAA1008; RGD1304646; RRP44
UniProt ID: (Human) Q9Y2L1, (Mouse) Q9CSH3
Entrez Gene ID: (Human) 22894, (Mouse) 72662, (Rat) 306103
Molecular Function:
RNA binding protein
endoribonuclease
exoribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.