Invitrogen
Manufacturer Code:PA567961
Catalog # PIPA567961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human DIO3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5DIII; 5DIII D3 DIOIII TXDI3 iodothyronine deiodinase placental type thyroxine deiodinase type III (selenoprotein) type 3 DI type 3 iodothyronine selenodeiodinase type III iodothyronine deiodinase type-III 5&prime deiodinase type-III 5&prime-deiodinase; deiodinase, iodothyronine type III; deiodinase, iodothyronine, type 3; DIOIII; iodothyronine deiodinase, placental type; Thyroxine 5-deiodinase; thyroxine deiodinase type III (selenoprotein); Type 3 DI; type 3 iodothyronine selenodeiodinase; Type III iodothyronine deiodinase; type-III 5' deiodinase; type-III 5'-deiodinase
Gene Aliases: 5DIII; D3; DIO3; DIOIII; ITDI3; TXDI3
UniProt ID: (Human) P55073, (Rat) P49897, (Mouse) Q91ZI8
Entrez Gene ID: (Human) 1735, (Rhesus monkey) 708027, (Rat) 29475, (Mouse) 107585
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.