Novus Biologicals
Manufacturer Code:NB11096414V
Catalog # NB11096414V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of rat DIO3 (within residues 1-100). [Swiss-Prot# P49897] |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5DIII; 5DIII D3 deiodinase iodothyronine type III DIOIII EC 1.97.1 EC 1.97.1.11 ITDI3 placental type thyroxine deiodinase type III (selenoprotein) Type 3 DI type 3 iodothyronine selenodeiodinase type III iodothyronine deiodinase type-III 5' deiodinase Type-III 5'-deiodinase; DIOIII; iodothyronine deiodinase, placental type; Thyroxine 5-deiodinase; thyroxine deiodinase type III (selenoprotein); Type 3 DI; type 3 iodothyronine selenodeiodinase; Type III iodothyronine deiodinase; type-III 5' deiodinase; type-III 5'-deiodinase
Gene Aliases: 5DIII; D3; DIO3; DIOIII; ITDI3; TXDI3
UniProt ID: (Human) P55073
Entrez Gene ID: (Human) 1735
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.