Novus Biologicals
Manufacturer Code:NBP29248220UL
Catalog # NB026835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 70-170 of human DIO3 (NP_001353.4). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5DIII; 5DIII D3 deiodinase iodothyronine type III DIOIII EC 1.97.1 EC 1.97.1.11 ITDI3 placental type thyroxine deiodinase type III (selenoprotein) Type 3 DI type 3 iodothyronine selenodeiodinase type III iodothyronine deiodinase type-III 5&prime deiodinase Type-III 5&prime-deiodinase; DIOIII; iodothyronine deiodinase, placental type; Thyroxine 5-deiodinase; thyroxine deiodinase type III (selenoprotein); Type 3 DI; type 3 iodothyronine selenodeiodinase; Type III iodothyronine deiodinase; type-III 5' deiodinase; type-III 5'-deiodinase
Gene Aliases: 5DIII; D3; DIO3; DIOIII; ITDI3; TXDI3
UniProt ID: (Human) P55073
Entrez Gene ID: (Human) 1735
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.