Invitrogen
Manufacturer Code:PA565489
Catalog # PIPA565489
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human DIO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5DII; deiodinase-2; deiodonase-2; DIO2 deiodinase iodothyronine type II 5DII D2 SelY TXDI2 thyroxine deiodinase type II type 2 iodothyronine deiodinase type II iodothyronine deiodinaseN: type-II 5&primedeiodinase; DIOII; thyroxine deiodinase, type II; Type 2 DI; type 2 iodothyronine deiodinase; Type II iodothyronine deiodinase; Type-II 5'-deiodinase; type-II 5'deiodinase
Gene Aliases: 5DII; D2; DIO2; DIOII; ITDI2; SelY; TXDI2
UniProt ID: (Human) Q92813
Entrez Gene ID: (Human) 1734
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.