Invitrogen
Manufacturer Code:PA5100474
Catalog # PIPA5100474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DIO2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5DII; 5DII AI324267 D2 deiodinase iodothyronine type 2 deiodinase iodothyronine type II deiodinase-2 deiodonase-2 Dio2 DIOII iodothyronine type II deiodinase Itdi2 SelY thyroxine deiodinase thyroxine deiodinase type II thyroxine type II TXDI2 Type 2 DI type 2 iodothyronine deiodinase Type II iodothyronine deiodinase type-II 5' deiodinase type-II 5'deiodinase type-II 5'-deiodinase; deiodinase, iodothyronine, type 2; deiodinase-2; deiodonase-2; DIOII; iodothyronine type II deiodinase; thyroxine deiodinase, type II; Type 2 DI; type 2 iodothyronine deiodinase; Type II iodothyronine deiodinase; type-II 5' deiodinase; Type-II 5'-deiodinase; type-II 5'deiodinase
Gene Aliases: 5DII; AI324267; D2; DIO2; DIOII; ITDI2; SelY; TXDI2
UniProt ID: (Human) Q92813, (Mouse) Q9Z1Y9, (Rat) P70551
Entrez Gene ID: (Human) 1734, (Mouse) 13371, (Rat) 65162
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.