Abnova
Manufacturer Code:PAB7542
Catalog # 89-133-350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human DIO2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of DIO2.
For Research Use Only
Protein Aliases: 5DII; D2; deiodinase-2; deiodonase-2; DIOII; SelY; thyroxine deiodinase, type II; TXDI2; Type 2 DI; type 2 iodothyronine deiodinase; Type II iodothyronine deiodinase; Type-II 5'-deiodinase; type-II 5'deiodinase
Gene Aliases: 5DII; D2; DIO2; DIOII; ITDI2; SelY; TXDI2
UniProt ID: (Human) Q92813
Entrez Gene ID: (Human) 1734
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.