Proteintech
Manufacturer Code:265131AP150UL
Catalog # 50-173-5361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DIO2 Fusion Protein Ag24119 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5DII; 5DII D2 DIO2 DIOII ITDI2 SelY TXDI2 Type 2 DI Type II 5 deiodinase; deiodinase-2; deiodonase-2; DIOII; iodothyronine type II deiodinase; thyroxine deiodinase, type II; Type 2 DI; type 2 iodothyronine deiodinase; Type II iodothyronine deiodinase; type-II 5' deiodinase; Type-II 5'-deiodinase; type-II 5'deiodinase
Gene Aliases: 5DII; AI324267; D2; DIO2; DIOII; ITDI2; SelY; TXDI2
UniProt ID: (Human) Q92813, (Mouse) Q9Z1Y9
Entrez Gene ID: (Human) 1734, (Mouse) 13371
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.