Abnova
Manufacturer Code:H00001733M01
Catalog # 89-021-302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E4 |
Immunogen | DIO1 (NP_000783.2 35 a.a. ~ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant DIO1.
For Research Use Only
Protein Aliases: 5DI; DIOI; iodothyronine deiodinase type 1; MGC130050; MGC130051; thyroxine 5'-deiodinase; thyroxine deiodinase type I (selenoprotein); TXDI1; Type 1 DI; Type I iodothyronine deiodinase; Type-I 5'-deiodinase
Gene Aliases: 5DI; DIO1; ITDI1; TXDI1
UniProt ID: (Human) P49895
Entrez Gene ID: (Human) 1733
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.