Invitrogen
Manufacturer Code:PA572504
Catalog # PIPA572504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 30-57 amino acids from the N-terminal region of human DIO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5DI; DIOI; iodothyronine deiodinase type 1; thyroxine 5'-deiodinase; thyroxine deiodinase type I (selenoprotein); Type 1 DI; Type I iodothyronine deiodinase; Type I iodothyronine deiodinase 5DI DIOI Type 1 DI Type-I 5&prime-deiodinase DIO1 ITDI1 TXDI1; Type-I 5'-deiodinase
Gene Aliases: 5DI; DIO1; ITDI1; TXDI1
UniProt ID: (Human) P49895
Entrez Gene ID: (Human) 1733
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.