Invitrogen
Manufacturer Code:PA5100139
Catalog # PIPA5100139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DIO1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5DI; 5DI D1 deiodinase iodothyronine type I deiodinase iodothyronine type I DIO1 DIOI hDido1 iodothyronine deiodinase type 1 ITDI1 thyroxine 5'-deiodinase thyroxine deiodinase thyroxine deiodinase type I (selenoprotein) thyroxine deiodinase type I TXDI1 Type 1 DI type I iodothyronine deiodinase Type-I 5'-deiodinase; DIOI; iodothyronine deiodinase type 1; thyroxine 5'-deiodinase; thyroxine deiodinase type I (selenoprotein); thyroxine deiodinase, type I; Type 1 DI; Type I iodothyronine deiodinase; Type-I 5'-deiodinase
Gene Aliases: 5DI; D1; DIO1; ITDI1; TXDI1
UniProt ID: (Human) P49895, (Mouse) Q61153, (Rat) P24389
Entrez Gene ID: (Human) 1733, (Mouse) 13370, (Rat) 25430
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.