Proteintech
Manufacturer Code:117901AP150UL
Catalog # 50-172-7457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DIO1 Fusion Protein Ag2410 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5DI; 5DI DIO1 DIOI ITDI1 TXDI1 Type 1 DI Type I 5 deiodinase; DIOI; iodothyronine deiodinase type 1; thyroxine 5'-deiodinase; thyroxine deiodinase type I (selenoprotein); thyroxine deiodinase, type I; Type 1 DI; Type I iodothyronine deiodinase; Type-I 5'-deiodinase
Gene Aliases: 5DI; D1; DIO1; ITDI1; TXDI1
UniProt ID: (Human) P49895, (Mouse) Q61153, (Rat) P24389
Entrez Gene ID: (Human) 1733, (Mouse) 13370, (Rat) 25430
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.