Novus Biologicals
Manufacturer Code:NBP297740AF532
Catalog # NB103209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human DIMT1L fragment. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase DIM1 DIM1 dimethyladenosine transferase 1-like DIM1 dimethyladenosine transferase 1-like (S. cerevisiae) DIMT1dimethyladenosine transferase EC 2.1.1 EC 2.1.1.14 EC 2.1.1.18318S rRNA dimethylase HSA9761 HUSSY5 N'-adenosyl(rRNA) dimethyltransferase probable dimethyladenosine transferase S-adenosylmethionine-6-N'; 18S rRNA dimethylase; DIM1 dimethyladenosine transferase 1 homolog; DIM1 dimethyladenosine transferase 1-like; dimethyladenosine transferase; Probable 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; Probable 18S rRNA dimethylase; Probable dimethyladenosine transferase; Probable S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase; S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase
Gene Aliases: DIM1; DIMT1; DIMT1L; HSA9761; HUSSY-05; HUSSY5
UniProt ID: (Human) Q9UNQ2
Entrez Gene ID: (Human) 27292
Molecular Function: RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.