Novus Biologicals
Manufacturer Code:NBP188480
Catalog # NBP188480
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LLPLDSHGVDFSRVRMWSLNNWSKYYRQTLLFGALQDAQINSVFNKYCVNMQGQVAVRNVPMTGSISHVLVQLPHVFQRMEAENLASVIDARFNFFVNKILPQYRDAVM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf107 chromosome 1 open reading frame 107 DEFFLJ41680 digestive organ expansion factor homolog digestive organ expansion factor homolog (zebrafish) digestive-organ expansion factor homolog DJ434O14.5 DKFZp686E19106 FLJ12704 MGC29875; Digestive organ expansion factor homolog; digestive-organ expansion factor homolog
Gene Aliases: C1orf107; DEF; DIEXF; DJ434O14.5; UTP25
UniProt ID: (Human) Q68CQ4
Entrez Gene ID: (Human) 27042
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.