Invitrogen
Manufacturer Code:PA131027
Catalog # PIPA131027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide VPLERSRSRHNGAISSK conjugated to KLH corresponding to amino acids 1085-1101 of Human DIA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: diaphanous homolog 2; diaphanous-related formin 2; Diaphanous-related formin-2; diaphorase-2; DRF2; mDia3 isoform; Protein diaphanous homolog 2; RP11-212B22.1 DIA DIA2 DRF2 FLJ11167 POF POF2 mDia3
Gene Aliases: DIA; DIA2; Dia3; Diap2; DIAPH2; DRF2; E430022I22Rik; POF; POF2
UniProt ID: (Human) Q8WX06, (Mouse) O70566
Entrez Gene ID: (Human) 1730, (Mouse) 54004
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.