Abnova
Manufacturer Code:PAB10201
Catalog # 89-114-689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 1085-1101 of human DIAPH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DIAPH2.
For Research Use Only
Protein Aliases: DIA; DIA2; diaphanous homolog 2; diaphanous-related formin 2; Diaphanous-related formin-2; diaphorase-2; DRF2; FLJ11167; POF; POF2; Protein diaphanous homolog 2
Gene Aliases: DIA; DIA2; DIAPH2; DRF2; POF; POF2
UniProt ID: (Human) Q8WX06
Entrez Gene ID: (Human) 1730
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.