Novus Biologicals
Manufacturer Code:NB600230UV
Catalog # NB600230UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide which represented a portion of human diaphanous homolog 1 encoded within exon 1 (LocusLink ID 1729). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DIA1 diaphanous homolog 1 (Drosophila) hDIA1 homolog) 1 LFHL1FLJ25265; diaphanous-related formin 1; Diaphanous-related formin-1; DRF1; Protein diaphanous homolog 1
Gene Aliases: DFNA1; DIA1; DIAP1; DIAPH1; DRF1; hDIA1; LFHL1; SCBMS
UniProt ID: (Human) O60610
Entrez Gene ID: (Human) 1729
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.