Novus Biologicals
Manufacturer Code:NBP250229V
Catalog # NBP250229V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | V78P3C10&asteriskD3 |
Immunogen | Peptide Sequence - RHNGAISSK |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DIA2 DIAdiaphanous (Drosophila homolog) 2 diaphanous homolog 2 (Drosophila) Diaphanous-related formin-2 diaphorase-2 DRF2 POF POF2FLJ11167 protein diaphanous homolog 2; diaphanous homolog 2; diaphanous-related formin 2; Diaphanous-related formin-2; diaphorase-2; DRF2; Protein diaphanous homolog 2
Gene Aliases: DIA; DIA2; DIAPH2; DRF2; POF; POF2
UniProt ID: (Human) Q8WX06
Entrez Gene ID: (Human) 1730
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.