Invitrogen
Manufacturer Code:PA541259
Catalog # PIPA541259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DHX9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase A; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9; DEAH (Asp-Glu-Ala-His) box helicase 9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DEAH (Asp-Glu-Ala-His) box polypeptide 9 DDX9 LKP NDH II NDHII RHA NDH2; DEAH box protein 9; DEAH-box helicase 9; DExH-box helicase 9; Leukophysin; LKP; mHEL-5; NDH II; Nuclear DNA helicase II; RHA; RNA helicase; RNA helicase A
Gene Aliases: AI326842; DDX9; DHX9; HEL-5; LKP; mHEL-5; NDH2; NDHII; RHA
UniProt ID: (Bovine) Q28141, (Human) Q08211, (Mouse) O70133
Entrez Gene ID: (Horse) 100055171, (Bovine) 281115, (Human) 1660, (Pig) 100514066, (Rabbit) 100354085, (Dog) 480032, (Mouse) 13211, (Rat) 304859
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.