Invitrogen
Manufacturer Code:PA578644
Catalog # PIPA578644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human DHX36. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent DNA/RNA helicase DHX36; ATP-dependent RNA helicase DHX36; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36; DEAD/H box polypeptide 36; DEAH (Asp-Glu-Ala-His) box polypeptide 36; DEAH (Asp-Glu-Ala-His) box polypeptide 36 DDX36 MLEL1 RHAU G4R1; DEAH box protein 36; DEAH-box protein 36; G4 resolvase-1; G4-resolvase 1; G4-resolvase-1; G4R1; MLE-like protein 1; probable ATP-dependent RNA helicase DHX36; RNA helicase associated with AU-rich element ARE; RNA helicase associated with AU-rich element protein
Gene Aliases: 2810407E23Rik; AI452301; AU022184; DDX36; DHX36; G4R1; KIAA1488; mKIAA1488; MLEL1; RHAU
UniProt ID: (Human) Q9H2U1, (Rat) D4A2Z8, (Mouse) Q8VHK9
Entrez Gene ID: (Human) 170506, (Rat) 310461, (Mouse) 72162
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.