Novus Biologicals
Manufacturer Code:NB200353
Catalog # NB200353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 950 and the C-terminus (residue 1008) of human DEAH (Asp-Glu-Ala-His) Box Polypeptide 36 using the numbering given in entry NP_065916.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent DNA/RNA helicase DHX36; DDX36 DEAH (Asp-Glu-Ala-His) box polypeptide 36 DEAH box protein 36 EC 3.6.1 EC 3.6.4.13 G4 resolvase-1 KIAA1488G4R1 MLEL1RHAUDEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36 MLE-like protein 1 probable ATP-dependent RNA helicase DHX36 RNA helicase associated with AU-rich element ARE; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36; DEAD/H box polypeptide 36; DEAH (Asp-Glu-Ala-His) box polypeptide 36; DEAH box protein 36; DEAH-box protein 36; G4 resolvase-1; G4-resolvase 1; G4-resolvase-1; G4R1; MLE-like protein 1; probable ATP-dependent RNA helicase DHX36; RNA helicase associated with AU-rich element ARE; RNA helicase associated with AU-rich element protein
Gene Aliases: DDX36; DHX36; G4R1; KIAA1488; MLEL1; RHAU
UniProt ID: (Human) Q9H2U1
Entrez Gene ID: (Human) 170506
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.