Novus Biologicals
Manufacturer Code:NBP213921
Catalog # NBP213921
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: EKGDIVVFLACEQDIEKVCETVYQGSNLNPDLGELVVVPLYPKEKCSLFK PLDETEKRCQVYQRRVVLTTSSGEFLIWSNSVRFVIDVGVERR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32 DEAD/H box 32 DEAD/H helicase-like protein 1 DEAH (Asp-Glu-Ala-His) box polypeptide 32 DEAH box protein 32 DHLP1DDX32DEAD/H helicase-like protein-1 EC 3.6.1 EC 3.6.4.13 FLJ10694 FLJ10889 huDDX32 putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32; DEAD/H box 32; DEAD/H helicase-like protein 1; DEAD/H helicase-like protein-1; DEAH (Asp-Glu-Ala-His) box polypeptide 32; DEAH box protein 32; DHLP1; HuDDX32; Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32
Gene Aliases: DDX32; DHLP1; DHX32
UniProt ID: (Human) Q7L7V1
Entrez Gene ID: (Human) 55760
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.