Novus Biologicals
Manufacturer Code:NBP29256120UL
Catalog # NB026993
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 906-1155 of human DHX30 (NP_055781.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DHX30; ATP-dependent RNA helicase DHX30 DDX30 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 30 DEAH (Asp-Glu-Ala-His) box polypeptide 30 DEAH box protein 30 EC 3.6.1 EC 3.6.4.13 FLJ11214 KIAA0890RETCOR putative ATP-dependent RNA helicase DHX30 retina co-repressor; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 30; DEAH (Asp-Glu-Ala-His) box helicase 30; DEAH (Asp-Glu-Ala-His) box polypeptide 30; DEAH box protein 30; retina co-repressor
Gene Aliases: DDX30; DHX30; KIAA0890; RETCOR
UniProt ID: (Human) Q7L2E3
Entrez Gene ID: (Human) 22907
Molecular Function:
RNA binding protein
RNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.