Invitrogen
Manufacturer Code:PA569451
Catalog # PIPA569451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse DHRS7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dehydrogenase/reductase (SDR family) member 7; Dehydrogenase/reductase SDR family member 7; Retinal short-chain dehydrogenase/reductase 4; retSDR4; SDR34C1 retDSR4 retSDR4 dehydrogenase/reductase SDR family member 7 retinal short-chain dehydrogenase/reductase 4 short chain dehydrogenase/reductase family 34C member 1; Short chain dehydrogenase/reductase family 34C member 1; short chain dehydrogenase/reductase family 34C, member 1; uncharacterized protein LOC299135
Gene Aliases: 2310016E22Rik; 5730564L20Rik; AW061210; CGI-86; DHRS7; DHRS7A; Retdsr4; RETSDR4; SDR34C1; UNQ285/PRO3448
UniProt ID: (Human) Q9Y394, (Mouse) Q9CXR1
Entrez Gene ID: (Dog) 480342, (Bovine) 514044, (Rabbit) 100340090, (Human) 51635, (Horse) 100060062, (Pig) 100152785, (Mouse) 66375, (Guinea pig) 100734266, (Rat) 299135
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.