Invitrogen
Manufacturer Code:PA556649
Catalog # PIPA556649
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DHRS7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dehydrogenase/reductase (SDR family) member 7; Dehydrogenase/reductase SDR family member 7; Retinal short-chain dehydrogenase/reductase 4; retSDR4; SDR34C1 retDSR4 retSDR4 dehydrogenase/reductase SDR family member 7 retinal short-chain dehydrogenase/reductase 4 short chain dehydrogenase/reductase family 34C member 1; Short chain dehydrogenase/reductase family 34C member 1; short chain dehydrogenase/reductase family 34C, member 1; uncharacterized protein LOC299135
Gene Aliases: CGI-86; DHRS7; DHRS7A; retDSR4; RETSDR4; SDR34C1; UNQ285/PRO3448
UniProt ID: (Human) Q9Y394
Entrez Gene ID: (Human) 51635, (Rat) 299135
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.