Abnova
Manufacturer Code:PAB27826
Catalog # 89-129-348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acid at internal region of human DHRS7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DHRS7.
For Research Use Only
Protein Aliases: CGI-86; dehydrogenase/reductase (SDR family) member 7; Dehydrogenase/reductase SDR family member 7; retDSR4; Retinal short-chain dehydrogenase/reductase 4; retSDR4; SDR34C1; Short chain dehydrogenase/reductase family 34C member 1; short chain dehydrogenase/reductase family 34C, member 1
Gene Aliases: CGI-86; DHRS7; DHRS7A; retDSR4; RETSDR4; SDR34C1; UNQ285/PRO3448
UniProt ID: (Human) Q9Y394
Entrez Gene ID: (Human) 51635
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.