Invitrogen
Manufacturer Code:PA5115119
Catalog # PIPA5115119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DHRS4 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI043103 AI790593 carbonyl reductase CR D14Ucla2 DCR-AKL dehydrogenase/reductase (SDR family) member 4 dehydrogenase/reductase (SDR family) member 4 like 2A dehydrogenase/reductase 4 dehydrogenase/reductase SDR family member 4 DHRS4 mouNRDR NADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase NADPH-dependent retinol dehydrogenase/reductase NADP-retinol dehydrogenase NDRD NRDR Peroxisomal short-chain alcohol dehydrogenase PH PSCD PHCR PSCD RRD SCAD-SRL SDR25C1 SDR25C2 SDR-SRL Short chain dehydrogenase/reductase family 25C member 2 short chain dehydrogenase/reductase family 25C member 1 short chain dehydrogenase/reductase family 25C member 2 Short-chain dehydrogenase/reductase family member 4 UNQ851/PRO1800; carbonyl reductase; CR; DCR-AKL; dehydrogenase/reductase (SDR family) member 4 like 2A; Dehydrogenase/reductase SDR family member 4; NADP-retinol dehydrogenase; NADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase; NADPH-dependent retinol dehydrogenase/reductase; NDRD; NRDR; Peroxisomal short-chain alcohol dehydrogenase; PHCR; PSCD; SCAD-SRL; Short chain dehydrogenase/reductase family 25C member 2; short chain dehydrogenase/reductase family 25C, member 1; short chain dehydrogenase/reductase family 25C, member 2; Short-chain dehydrogenase/reductase family member 4
Gene Aliases: AI043103; AI790593; CR; D14Ucla2; DHRS4; mouNRDR; NDRD; NRDR; PHCR; PSCD; RRD; SCAD-SRL; SDR-SRL; SDR25C1; SDR25C2; UNQ851/PRO1800
UniProt ID: (Human) Q9BTZ2, (Rat) Q8VID1, (Mouse) Q99LB2
Entrez Gene ID: (Human) 10901, (Rat) 266686, (Mouse) 28200
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.