Novus Biologicals
Manufacturer Code:NBP298675FR
Catalog # NB105124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human DHRS4 fragment. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CR; CR dehydrogenase/reductase (SDR family) member 4 dehydrogenase/reductase (SDR family) member 4 like 2A dehydrogenase/reductase SDR family member 4 EC 1.1.1 EC 1.1.1.184 FLJ11008 humNRDR NADP-dependent retinol dehydrogenase NRDRNADPH-dependent retinol dehydrogenase/reductase Peroxisomal short-chain alcohol dehydrogenase PHCR PSCD SCAD-SRLNADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase SDR25C1 SDR25C2 SDR-SRL short chain dehydrogenase/reductase family 25C member 1 short chain dehydrogenase/reductase family 25C member 2 Short-chain dehydrogenase/reductase family member 4; dehydrogenase/reductase (SDR family) member 4 like 2A; Dehydrogenase/reductase SDR family member 4; NADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase; NADPH-dependent retinol dehydrogenase/reductase; NRDR; Peroxisomal short-chain alcohol dehydrogenase; PSCD; SCAD-SRL; Short chain dehydrogenase/reductase family 25C member 2; short chain dehydrogenase/reductase family 25C, member 1; short chain dehydrogenase/reductase family 25C, member 2; Short-chain dehydrogenase/reductase family member 4
Gene Aliases: CR; DHRS4; NRDR; PHCR; PSCD; SCAD-SRL; SDR-SRL; SDR25C1; SDR25C2; UNQ851/PRO1800
UniProt ID: (Human) Q9BTZ2
Entrez Gene ID: (Human) 10901
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.