Invitrogen
Manufacturer Code:PA539129
Catalog # PIPA539129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human DHRS11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-hydroxysteroid dehydrogenase; 3-beta-hydroxysteroid 3-dehydrogenase; dehydrogenase/reductase (SDR family) member 11; Dehydrogenase/reductase SDR family member 11; Estradiol 17-beta-dehydrogenase; inactive dehydrogenase/reductase isoform; short chain dehydrogenase/reductase family 24C, member 1; Short-chain dehydrogenase/reductase family 24C member 1
Gene Aliases: ARPG836; DHRS11; SDR24C1; spDHRS11; UNQ836/PRO1774
UniProt ID: (Human) Q6UWP2, (Mouse) Q3U0B3
Entrez Gene ID: (Human) 79154, (Mouse) 192970
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.