OriGene
Manufacturer Code:TA812489
Catalog # 50-168-4568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4C12 |
Immunogen | Full length human recombit protein of human DHRS1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dehydrogenase/reductase (SDR family) member 1; Dehydrogenase/reductase SDR family member 1; SDR19C1; Short chain dehydrogenase/reductase family 19C member 1; short chain dehydrogenase/reductase family 19C, member 1
Gene Aliases: DHRS1; SDR19C1
UniProt ID: (Human) Q96LJ7
Entrez Gene ID: (Human) 115817
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.