Invitrogen
Manufacturer Code:PA568164
Catalog # PIPA568164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human DHRS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dehydrogenase/reductase (SDR family) member 1; Dehydrogenase/reductase SDR family member 1; Dehydrogenase/reductase SDR family member 1 11-- DHRS1; Short chain dehydrogenase/reductase family 19C member 1; short chain dehydrogenase/reductase family 19C, member 1
Gene Aliases: 1110029G07Rik; AW112170; D14Ertd484e; DHRS1; SDR19C1
UniProt ID: (Human) Q96LJ7, (Mouse) Q99L04
Entrez Gene ID: (Human) 115817, (Mouse) 52585, (Rat) 290234
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.